Tisk…

Anti-CEBPA Mouse monoclonal antibody unconjugated [clone: 4E10]

Dodavatel: Abcam
ABCAAB128162-100EA 26320 CZK
ABCAAB128162-100
Anti-CEBPA Mouse monoclonal antibody unconjugated [clone: 4E10]
Protilátky
Type: Primary
Antigen: CEBPA (CCAAT/enhancer binding protein (C/EBP), alpha)
Clonality: monoclonal
Clone: 40000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR