Tisk…

Anti-CBFA2T2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB128164-100EA 24390 CZK
ABCAAB128164-100
Anti-CBFA2T2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CBFA2T2 (core-binding factor, runt domain, alpha subunit 2; translocated to, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR