Tisk…

Anti-TRIP13 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB128171-100EA 25740 CZK
ABCAAB128171-100
Anti-TRIP13 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRIP13 (thyroid hormone receptor interactor 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR