Tisk…
Anti-PYGO1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB128174-100EA 28190 CZK
ABCAAB128174-100
Anti-PYGO1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PYGO1 (pygopus family PHD finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human