Tisk…

Anti-ZCH11 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB128180-100EA 25590 CZK
ABCAAB128180-100
Anti-ZCH11 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ZCH11
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR