Tisk…

Anti-ABCA13 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB128248-100EA 24420 CZK
ABCAAB128248-100
Anti-ABCA13 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ABCA13 (ATP-binding cassette, sub-family A (ABC1), member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR