Tisk…
Anti-ACVR2B Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB128544-100EA 25680 CZK
ABCAAB128544-100
Anti-ACVR2B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ACVR2B (activin A receptor, type IIB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human