Tisk…

Anti-PCDH17 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB128815-100EA 25590 CZK
ABCAAB128815-100
Anti-PCDH17 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PCDH17 (protocadherin 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR