Tisk…
Anti-LAMB3 Rabbit Monoclonal Antibody [clone: EPR7524]
Dodavatel: Abcam
ABCAAB128864-100EA 24120 CZK
ABCAAB128864-100
Anti-LAMB3 Rabbit Monoclonal Antibody [clone: EPR7524]
Protilátky
Type: Primary
Antigen: LAMB3 (laminin, beta 3)
Clonality: monoclonal
Clone: EPR7524
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat