Tisk…

Anti-FASN Rabbit monoclonal antibody unconjugated [clone: EPR7466]

Dodavatel: Abcam
ABCAAB128870-100EA 29290 CZK
ABCAAB128870-100
Anti-FASN Rabbit monoclonal antibody unconjugated [clone: EPR7466]
Protilátky
Type: Primary
Antigen: FASN (fatty acid synthase)
Clonality: monoclonal
Clone: EPR7466
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR