Tisk…

Anti-SMC3 Rabbit Monoclonal Antibody [clone: EPR7984]

Dodavatel: Abcam
ABCAAB128919-100EA 26320 CZK
ABCAAB128919-100
Anti-SMC3 Rabbit Monoclonal Antibody [clone: EPR7984]
Protilátky
Type: Primary
Antigen: SMC3 (structural maintenance of chromosomes 3)
Clonality: monoclonal
Clone: EPR7984
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR