Tisk…

Anti-MCM3 Rabbit Monoclonal Antibody [clone: EPR7080]

Dodavatel: Abcam
ABCAAB128923-100EA 26170 CZK
ABCAAB128923-100
Anti-MCM3 Rabbit Monoclonal Antibody [clone: EPR7080]
Protilátky
Type: Primary
Antigen: MCM3 (minichromosome maintenance complex component 3)
Clonality: monoclonal
Clone: EPR7080
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR