Tisk…

Anti-PRDX3 Rabbit Monoclonal Antibody [clone: EPR8115]

Dodavatel: Abcam
ABCAAB128953-100EA 28830 CZK
ABCAAB128953-100
Anti-PRDX3 Rabbit Monoclonal Antibody [clone: EPR8115]
Protilátky
Type: Primary
Antigen: PRDX3 (peroxiredoxin 3)
Clonality: monoclonal
Clone: EPR8115
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR