Tisk…
Anti-GLB1 Rabbit Monoclonal Antibody [clone: EPR8250]
Dodavatel: Abcam
ABCAAB128993-100EA 25740 CZK
ABCAAB128993-100
Anti-GLB1 Rabbit Monoclonal Antibody [clone: EPR8250]
Protilátky
Type: Primary
Antigen: GLB1 (galactosidase, beta 1)
Clonality: monoclonal
Clone: EPR8250
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat