Tisk…

Anti-FTCD Rabbit Monoclonal Antibody [clone: EPR7909]

Dodavatel: Abcam
ABCAAB129005-100EA 26140 CZK
ABCAAB129005-100
Anti-FTCD Rabbit Monoclonal Antibody [clone: EPR7909]
Protilátky
Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: monoclonal
Clone: EPR7909
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR