Tisk…

Anti-MVD Rabbit Monoclonal Antibody [clone: EPR6559]

Dodavatel: Abcam
ABCAAB129061-100EA 25560 CZK
ABCAAB129061-100
Anti-MVD Rabbit Monoclonal Antibody [clone: EPR6559]
Protilátky
Type: Primary
Antigen: MVD (mevalonate (diphospho) decarboxylase)
Clonality: monoclonal
Clone: EPR6559
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR