Tisk…

Anti-ACO2 Rabbit Monoclonal Antibody [clone: EPR8282]

Dodavatel: Abcam
ABCAAB129069-100EA 27150 CZK
ABCAAB129069-100
Anti-ACO2 Rabbit Monoclonal Antibody [clone: EPR8282]
Protilátky
Type: Primary
Antigen: ACO2 (aconitase 2, mitochondrial)
Clonality: monoclonal
Clone: EPR8282
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR