Tisk…
Anti-IGF2BP2 Rabbit Monoclonal Antibody [clone: EPR6740]
Dodavatel: Abcam
ABCAAB129071-100EA 27820 CZK
ABCAAB129071-100
Anti-IGF2BP2 Rabbit Monoclonal Antibody [clone: EPR6740]
Protilátky
Type: Primary
Antigen: IGF2BP2 (insulin-like growth factor 2 mRNA binding protein 2)
Clonality: monoclonal
Clone: EPR6740
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat