Tisk…

Anti-LTK Rabbit Monoclonal Antibody [clone: EPR5175]

Dodavatel: Abcam
ABCAAB129155-100EA 23900 CZK
ABCAAB129155-100
Anti-LTK Rabbit Monoclonal Antibody [clone: EPR5175]
Protilátky
Type: Primary
Antigen: LTK (leukocyte receptor tyrosine kinase)
Clonality: monoclonal
Clone: EPR5175
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR