Tisk…

Anti-ERGI3 Rabbit Monoclonal Antibody [clone: EPR8141]

Dodavatel: Abcam
ABCAAB129179-100EA 24060 CZK
ABCAAB129179-100
Anti-ERGI3 Rabbit Monoclonal Antibody [clone: EPR8141]
Protilátky
Type: Primary
Antigen: ERGI3
Clonality: monoclonal
Clone: EPR8141
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR