Tisk…

Anti-KCNJ3 Rabbit Monoclonal Antibody [clone: EPR6363]

Dodavatel: Abcam
ABCAAB129182-100EA 26940 CZK
ABCAAB129182-100
Anti-KCNJ3 Rabbit Monoclonal Antibody [clone: EPR6363]
Protilátky
Type: Primary
Antigen: KCNJ3 (potassium inwardly-rectifying channel, subfamily J, member 3)
Clonality: monoclonal
Clone: EPR6363
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR