Tisk…

Anti-VPS45 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB129298-50EA 26140 CZK
ABCAAB129298-50
Anti-VPS45 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: VPS45 (vacuolar protein sorting 45 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR