Tisk…
Anti-CRKL Goat polyclonal antibody unconjugated
Dodavatel: Abcam
ABCAAB129318-50EA 25740 CZK
ABCAAB129318-50
Anti-CRKL Goat polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: CRKL (v-crk avian sarcoma virus CT10 oncogene homolog-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human