Tisk…

Anti-EPM2A Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB129321-50EA 24420 CZK
ABCAAB129321-50
Anti-EPM2A Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR