Tisk…

Anti-ALDOB Mouse Monoclonal Antibody [clone: 5E2AD2]

Dodavatel: Abcam
ABCAAB129728-100EA 26050 CZK
ABCAAB129728-100
Anti-ALDOB Mouse Monoclonal Antibody [clone: 5E2AD2]
Protilátky
Type: Primary
Antigen: ALDOB (aldolase B, fructose-bisphosphate)
Clonality: monoclonal
Clone: 5E2AD2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR