Tisk…

Anti-ATP5G3 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB129742-100EA 24420 CZK
ABCAAB129742-100
Anti-ATP5G3 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: ATP5G3 (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR