Tisk…

Anti-RFTN1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB129765-100EA 24420 CZK
ABCAAB129765-100
Anti-RFTN1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RFTN1 (raftlin, lipid raft linker 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR