Tisk…

Anti-TP53INP2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB129784-100EA 26140 CZK
ABCAAB129784-100
Anti-TP53INP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TP53INP2 (tumor protein p53 inducible nuclear protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR