Tisk…

Anti-OSR2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB129897-100EA 24420 CZK
ABCAAB129897-100
Anti-OSR2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: OSR2 (odd-skipped related transciption factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR