Tisk…
Anti-TMEM185A Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB130101-100EA 24420 CZK
ABCAAB130101-100
Anti-TMEM185A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TMEM185A (transmembrane protein 185A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human