Tisk…
Anti-DAXX Mouse monoclonal antibody unconjugated [clone: 7A11]
Dodavatel: Abcam
ABCAAB130198-100EA 25310 CZK
ABCAAB130198-100
Anti-DAXX Mouse monoclonal antibody unconjugated [clone: 7A11]
Protilátky
Type: Primary
Antigen: DAXX (death-domain associated protein)
Clonality: monoclonal
Clone: 7A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human