Tisk…

Anti-BAP1 Mouse monoclonal antibody unconjugated [clone: BP-1]

Dodavatel: Abcam
ABCAAB130202-100EA 25740 CZK
ABCAAB130202-100
Anti-BAP1 Mouse monoclonal antibody unconjugated [clone: BP-1]
Protilátky
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: BP-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR