Tisk…

Anti-Macrophage L1 protein Calprotectin Mouse Monoclonal Antibody [clone: B314.1 (MAC 387)]

Dodavatel: Abcam
ABCAAB130234-500EA 26140 CZK
ABCAAB130234-500
Anti-Macrophage L1 protein Calprotectin Mouse Monoclonal Antibody [clone: B314.1 (MAC 387)]
Protilátky
Type: Primary
Antigen: Macrophage L1 protein Calprotectin
Clonality: monoclonal
Clone: B314.1 (MAC 387)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR