Tisk…

Anti-IgG2a Goat Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)

Dodavatel: Abcam
ABCAAB130787-125EA 12050 CZK
ABCAAB130787-125
Anti-IgG2a Goat Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
Protilátky
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR