Tisk…

Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: Abcam
ABCAAB130805-125EA 13790 CZK
ABCAAB130805-125
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR