Tisk…

Anti-TUB 1 Rabbit Monoclonal Antibody [clone: EPR8241]

Dodavatel: Abcam
ABCAAB131034-100EA 23440 CZK
ABCAAB131034-100
Anti-TUB 1 Rabbit Monoclonal Antibody [clone: EPR8241]
Protilátky
Type: Primary
Antigen: TUB 1
Clonality: monoclonal
Clone: EPR8241
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR