Tisk…

Anti-HGD Rabbit Monoclonal Antibody [clone: EPR7874]

Dodavatel: Abcam
ABCAAB131035-100EA 23870 CZK
ABCAAB131035-100
Anti-HGD Rabbit Monoclonal Antibody [clone: EPR7874]
Protilátky
Type: Primary
Antigen: HGD (homogentisate 1,2-dioxygenase)
Clonality: monoclonal
Clone: EPR7874
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR