Tisk…

Anti-LTB4R Rabbit Monoclonal Antibody [clone: EPR7113]

Dodavatel: Abcam
ABCAAB131041-100EA 24120 CZK
ABCAAB131041-100
Anti-LTB4R Rabbit Monoclonal Antibody [clone: EPR7113]
Protilátky
Type: Primary
Antigen: LTB4R (leukotriene B4 receptor)
Clonality: monoclonal
Clone: EPR7113
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR