Tisk…

Anti-ERBB2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB131102-100EA 26110 CZK
ABCAAB131102-100
Anti-ERBB2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR