Tisk…

Anti-EEF2 Mouse monoclonal antibody [clone: 4B3-G7-H5]

Dodavatel: Abcam
ABCAAB131202-100EA 23930 CZK
ABCAAB131202-100
Anti-EEF2 Mouse monoclonal antibody [clone: 4B3-G7-H5]
Type: Primary
Antigen: EEF2 (eukaryotic translation elongation factor 2)
Clonality: monoclonal
Clone: 4B3-G7-H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR