Tisk…

Anti-CLDN5 Rabbit monoclonal antibody unconjugated [clone: EPR7583]

Dodavatel: Abcam
ABCAAB131259-100EA 28620 CZK
ABCAAB131259-100
Anti-CLDN5 Rabbit monoclonal antibody unconjugated [clone: EPR7583]
Protilátky
Type: Primary
Antigen: CLDN5 (claudin 5)
Clonality: monoclonal
Clone: EPR7583
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR