Tisk…

Anti-ARF6 Rabbit Monoclonal Antibody [clone: EPR8357]

Dodavatel: Abcam
ABCAAB131261-100EA 28830 CZK
ABCAAB131261-100
Anti-ARF6 Rabbit Monoclonal Antibody [clone: EPR8357]
Protilátky
Type: Primary
Antigen: ARF6 (ADP-ribosylation factor 6)
Clonality: monoclonal
Clone: EPR8357
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR