Tisk…

Anti-AK3 Rabbit Monoclonal Antibody [clone: EPR7678]

Dodavatel: Abcam
ABCAAB131327-100EA 24120 CZK
ABCAAB131327-100
Anti-AK3 Rabbit Monoclonal Antibody [clone: EPR7678]
Protilátky
Type: Primary
Antigen: AK3 (adenylate kinase 3)
Clonality: monoclonal
Clone: EPR7678
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR