Tisk…

Anti-AKR1C1 Mouse Monoclonal Antibody [clone: 4B6AF3]

Dodavatel: Abcam
ABCAAB131375-100EA 25800 CZK
ABCAAB131375-100
Anti-AKR1C1 Mouse Monoclonal Antibody [clone: 4B6AF3]
Protilátky
Type: Primary
Antigen: AKR1C1 (aldo-keto reductase family 1, member C1)
Clonality: monoclonal
Clone: 4B6AF3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR