Tisk…

Anti-Estradiol Mouse Monoclonal Antibody [clone: B42]

Dodavatel: Abcam
ABCAAB131413-500EA 25560 CZK
ABCAAB131413-500
Anti-Estradiol Mouse Monoclonal Antibody [clone: B42]
Protilátky
Type: Primary
Antigen: Estradiol
Clonality: monoclonal
Clone: B42
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR