Tisk…

Anti-BCL2L11 Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB131477-100EA 25740 CZK
ABCAAB131477-100
Anti-BCL2L11 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: BCL2L11 (BCL2-like 11 (apoptosis facilitator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR