Tisk…
Anti-STMN1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB131481-100EA 26320 CZK
ABCAAB131481-100
Anti-STMN1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: STMN1 (stathmin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat