Tisk…

Anti-PTK2B Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB131543-100EA 26410 CZK
ABCAAB131543-100
Anti-PTK2B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PTK2B (protein tyrosine kinase 2 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR