Tisk…

Anti-SMAD1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB131550-100EA 25740 CZK
ABCAAB131550-100
Anti-SMAD1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SMAD1 (SMAD family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR