Tisk…
Anti-EZR Rabbit monoclonal antibody unconjugated [clone: EPR803]
Dodavatel: Abcam
ABCAAB133297-100EA 26140 CZK
ABCAAB133297-100
Anti-EZR Rabbit monoclonal antibody unconjugated [clone: EPR803]
Protilátky
Type: Primary
Antigen: EZR (ezrin)
Clonality: monoclonal
Clone: EPR803
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat